Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FABP6 Recombinant Rabbit Monoclonal Antibody (009)
GREENER_CHOICE

Catalog No. PIMA530405
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530405 Supplier Invitrogen™ Supplier No. MA530405
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human FABP6/ILBP/I-BABP.

This gene encodes the ileal fatty acid binding protein. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABP6 and FABP1 (the liver fatty acid binding protein) are also able to bind bile acids. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism. Transcript variants generated by alternate transcription promoters and/or alternate splicing have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FABP6
Applications ELISA
Classification Recombinant Monoclonal
Clone 009
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene FABP6
Gene Accession No. P51161
Gene Alias FABP6; fatty acid binding protein 6; fatty acid binding protein 6, ileal; fatty acid binding protein 6, ileal (gastrotropin); fatty acid-binding protein 6; Gastrotropin; GT; I-15P; I-BABP; I-BALB; I-BAP; ILBP; ILBP3; ileal bile acid binding protein; ileal bile acid-binding protein; ileal lipid-binding protein; Illbp; illeal lipid-binding protein; intestinal 15 kDa protein; Intestinal bile acid-binding protein
Gene Symbols FABP6
Host Species Rabbit
Immunogen Recombinant Human FABP6/ILBP/I-BABP Protein (Met1-Ala128).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2172
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.