Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP7/B-FABP Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $427.50

Specifications

Antigen FABP7/B-FABP
Concentration 1.0 mg/mL
Dilution Western Blot 2-4ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227073SS
SDP
View Documents
Novus Biologicals
NBP227073SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP227073
SDP
View Documents
Novus Biologicals
NBP227073
0.1 mg
Each for $427.50
Only null left
Add to Cart
 
Description

Description

FABP7/B-FABP Polyclonal specifically detects FABP7/B-FABP in Human, Mouse, Bovine, Equine, Opossum, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

FABP7/B-FABP
Western Blot 2-4ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml
Polyclonal
Purified
RUO
Human, Mouse, Bovine, Equine, Opossum, Primate
O15540
2173
A portion of amino acids 80-132 of human FABP7 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1.0 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cell Cycle and Replication, Stem Cells
0.1 ml PBS with 0.05% Sodium Azide
B-FABPDKFZp547J2313, BLBPMRG, Brain lipid-binding protein, Brain-type fatty acid-binding protein, FABPBbrain lipid binding protein, fatty acid binding protein 7, brain, Fatty acid-binding protein 7, fatty acid-binding protein, brain, Mammary-derived growth inhibitor related, mammary-derived growth inhibitor-related
FABP7
IgG
Protein A purified
The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, monkey, dog, cow and horse, and 94% homologous in xenopus, chicken, rat and pufferfish, and 88% homologous in mouse.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.