Learn More
FABP7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
This antibody was raised against the full-length human FABP7 and may cross-react with other FABPs.
Specifications
Specifications
| Antigen | FABP7 |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.33 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | FABP7 |
| Gene Accession No. | O15540, P51880, P55051 |
| Gene Alias | B FABP, BLBP, Brain lipid binding protein, FABP7, Fatty acid binding protein 7, MRG |
| Gene Symbols | Fabp7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.