Learn More
FABP7 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 148361AP150UL

Description
This antibody specifically recognizes FABP7 and not cross-react with other FABPs.
FABP7 was initially isolated from a human fetal brain cDNA library and whose mRNA was expressed in adult brain and muscle tissues at low levels. The protein encoded by this gene is a member of the fatty acid binding protein (FABPs) family, a group of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABPs are thought to play roles in fatty acid uptake, transport, and metabolism. FABP7 is a downstream gene of the Pax6 transcription factor and has been suggested to be essential for the maintece of neuroepithelial cells during early cortical development. More recently, FABP7 was found to be frequently expressed in melanomas. Down-regulation of FABP7 through RNAi expression could reduce in vitro cell proliferation and Matrigel invasion, suggesting that FABP7 may be a potential target for the development of diagnostic and therapeutic tools.Specifications
FABP7 | |
Polyclonal | |
Unconjugated | |
FABP7 | |
B FABP, BLBP, Brain lipid binding protein, FABP7, FABPB, Fatty acid binding protein 7, MRG | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2173 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15540 | |
FABP7 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.