Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP8/M-FABP/Myelin P2 Protein Antibody (3H3F4D5), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306650F
Description
FABP8/M-FABP/Myelin P2 Protein Monoclonal antibody specifically detects FABP8/M-FABP/Myelin P2 Protein in Human samples. It is validated for ELISASpecifications
FABP8/M-FABP/Myelin P2 Protein | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
3H3F4D5 | |
ELISA | |
M-FABP, MP2, myelin P2 protein, P2, peripheral myelin protein 2FABP8 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human FABP8/M-FABP/Myelin P2 Protein (NP_002668.1; Ser 2-Val 132). T | |
0.1 mL | |
Cardiovascular Biology, Endocrinology, Signal Transduction | |
5375 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction