Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP8/M-FABP/Myelin P2 Protein Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299794JF646
Description
FABP8/M-FABP/Myelin P2 Protein Polyclonal antibody specifically detects FABP8/M-FABP/Myelin P2 Protein in Human samples. It is validated for ELISASpecifications
FABP8/M-FABP/Myelin P2 Protein | |
Polyclonal | |
ELISA | |
M-FABP, MP2, myelin P2 protein, P2, peripheral myelin protein 2FABP8 | |
Produced in rabbits immunized with purified, recombinant Human FABP8/M-FABP/Myelin P2 Protein (Accession#: NP_002668.1; Ser 2-Val 132) | |
0.1 mL | |
Cardiovascular Biology, Endocrinology, Signal Transduction | |
5375 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction