Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FABP8 Monoclonal Antibody (3H3F4D5)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531128
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human PMP2/FABP8.
The protein encoded by this gene localizes to myelin sheaths of the peripheral nervous system. The encoded protein can bind both the membrane layers of the sheaths and monomeric lipids, and is thought to provide stability to the sheath. A defect in this gene was shown to be a cause of dominant demyelinating CMT neuropathy.
Specifications
FABP8 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P02689 | |
PMP2 | |
Recombinant Human PMP2 protein (Ser2-Val132). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
3H3F4D5 | |
Unconjugated | |
PMP2 | |
FABP8; M FABP8; M-FABP; MP2; myelin P2 protein; P2; Peripheral myelin protein 2; PMP2 | |
Mouse | |
Protein A | |
RUO | |
5375 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction