Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FABP8 Monoclonal Antibody (631306)
GREENER_CHOICE

Catalog No. PIMA524283
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524283 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524283 Supplier Invitrogen™ Supplier No. MA524283
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In direct ELISAs, no cross-reactivity with recombinant human FABP1, 2, 3, 4, 5, 6, 7, or 9 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

The protein encoded by this gene localizes to myelin sheaths of the peripheral nervous system. The encoded protein can bind both the membrane layers of the sheaths and monomeric lipids, and is thought to provide stability to the sheath. A defect in this gene was shown to be a cause of dominant demyelinating CMT neuropathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FABP8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 631306
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene PMP2
Gene Accession No. P02689
Gene Alias FABP8; M FABP8; M-FABP; MP2; myelin P2 protein; P2; Peripheral myelin protein 2; PMP2
Gene Symbols PMP2
Host Species Mouse
Immunogen E. coli-derived recombinant human FABP8/M-FABP Ser2-Val132.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5375
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.