Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FABP8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597162
Description
Immunogen sequence: MSNKFLGTWK LVSSENFDDY MKALGVGLAT RKLGNLAKPT VIISKKGDII TIRTESTFKN TEISFKLGQE FEETTADNRK TKSIVTLQRG SLNQVQRWDG KETTIKRKLV NGKMVAECKM KGVVCTRIYE KV; Positive Samples: Mouse small intestine; Cellular Location: Cytoplasm.
The protein encoded by this gene localizes to myelin sheaths of the peripheral nervous system. The encoded protein can bind both the membrane layers of the sheaths and monomeric lipids, and is thought to provide stability to the sheath. A defect in this gene was shown to be a cause of dominant demyelinating CMT neuropathy.
Specifications
FABP8 | |
Polyclonal | |
Unconjugated | |
PMP2 | |
FABP8; M FABP8; M-FABP; MP2; myelin P2 protein; P2; Peripheral myelin protein 2; PMP2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
18857 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
4.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P24526 | |
PMP2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-132 of human PMP2 (NP_0026681). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction