Learn More
Invitrogen™ FABP9 Monoclonal Antibody (324926)

Description
In direct ELISAs, 25% cross-reactivity with recombinant human (rh) FABP6 and rhFABP7 is observed and no cross-reactivity with recombinant mouse (rm) FABP4, rmFABP5, recombinant rat (rr) FABP1, rrFABP2, or rhFABP3. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | FABP9 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 324926 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Fabp9 |
| Gene Accession No. | O08716 |
| Gene Alias | 15 kDa perforatorial protein; 1700007P10Rik; FABP9; FABP-9; fatty acid binding protein 9; fatty acid binding protein 9, testis; fatty acid-binding protein 9; lipid-binding protein; PERF; PERF 15; Perf15; testis fatty acid binding protein; testis lipid binding protein; testis lipid-binding protein; Testis-type fatty acid-binding protein; T-FABP; TLBP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.