Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FABP9 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. FEPA5145387
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145387 200 μL
FEPA5145388 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145387 Supplier Invitrogen™ Supplier No. PA5145387
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Fatty acid-binding protein 9 (FABP9), also known as testis FABP, is a gene located on chromosome 8q21.1 that encodes a protein primarily expressed in the testis, where it plays a critical role in fatty acid metabolism and transport. FABP9 is part of a larger family of fatty acid-binding proteins that facilitate the intracellular movement and storage of lipids, impacting cellular signaling and energy homeostasis. Structurally, FABP9 is adapted to bind long-chain fatty acids and other hydrophobic molecules, contributing to the regulation of lipid dynamics within spermatogenic cells. The expression of FABP9 is crucial during spermatogenesis, influencing sperm maturation and fertility. Beyond its reproductive role, variations in FABP9 have been studied in relation to metabolic disorders, linking its lipid regulatory functions to broader physiological processes. Research continues to explore FABP9's potential involvement in systemic lipid metabolism and its implications for diseases associated with lipid dysregulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FABP9
Applications Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene Fabp9
Gene Accession No. O08716, P55054, Q0Z7S8
Gene Alias 15 kDa perforatorial protein; 1700007P10Rik; FABP9; FABP-9; fatty acid binding protein 9; fatty acid binding protein 9, testis; fatty acid-binding protein 9; lipid-binding protein; PERF; PERF 15; Perf15; testis fatty acid binding protein; testis lipid binding protein; testis lipid-binding protein; Testis-type fatty acid-binding protein; T-FABP; TLBP
Gene Symbols Fabp9
Host Species Rabbit
Immunogen Recombinant Human FABP9 protein, Leu1-Val132 (Accession #Q0Z7S8).
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21884, 646480, 64822
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.