Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP9/T-FABP Antibody (13F9), Alexa Fluor™ 350, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP231200AF350

Catalog No. NP23120A350


Only null left
Add to Cart

Description

Description

FABP9/T-FABP Monoclonal specifically detects FABP9/T-FABP in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

FABP9/T-FABP
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
FABP9
Recombinant human FABP9 (1-132aa) purified from E. coli
0.1 mL
Primary
Human
IgG1 κ
Western Blot, ELISA
13F9
Western Blot, ELISA
FABP-9, Fatty Acid Binding Protein 9, Testis, Fatty Acid-Binding Protein 9, Lipid-Binding Protein, PERF, PERF15, Testis Lipid Binding Protein, Testis Lipid-Binding Protein, Testis-Type Fatty Acid-Binding Protein, T-FABP, TLBP
Mouse
Protein A purified
Developmental Biology, metabolism
646480.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.