Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP9/T-FABP Antibody (13F9), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP231200G
Description
FABP9/T-FABP Monoclonal specifically detects FABP9/T-FABP in Human samples. It is validated for Western Blot, ELISA.Specifications
| FABP9/T-FABP | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FABP9 | |
| Recombinant human FABP9 (1-132aa) purified from E. coli | |
| 0.1 mL | |
| Developmental Biology, metabolism | |
| 646480 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 13F9 | |
| Western Blot, ELISA | |
| FABP-9, Fatty Acid Binding Protein 9, Testis, Fatty Acid-Binding Protein 9, Lipid-Binding Protein, PERF, PERF15, Testis Lipid Binding Protein, Testis Lipid-Binding Protein, Testis-Type Fatty Acid-Binding Protein, T-FABP, TLBP | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Anti-human FABP9 mAb, clone 13F9, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human FABP9 protein. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction