Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FACA/FANCA Rabbit anti-Human, Clone: 4Q1Q8, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP316818100UL
This item is not returnable.
View return policy
Description
FACA/FANCA Monoclonal antibody specifically detects FACA/FANCA in Human samples. It is validated for Western BlotSpecifications
| FACA/FANCA | |
| Monoclonal | |
| Unconjugated | |
| FA, FA1, FAAMGC75158, FACAFanconi anemia group A protein, FAH, FA-H, FANCH, Fanconi anemia, complementation group A, Fanconi anemia, complementation group H, Fanconi anemia, type 1, Protein FACA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 2-104 of human FACA/FANCA (O15360). | |
| 100 μg | |
| Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
| 2175 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 4Q1Q8 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction