Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor 10 heavy chain Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989116
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9116 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9116 Supplier Bioss Supplier No. BS9501R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen Factor 10 heavy chain
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene F10
Gene Accession No. P00742
Gene Alias Activated factor Xa heavy chain; BOS_12710; Coagulation factor X; F10; F10a; Factor X heavy chain; Factor X light chain; factor Xa; FX; FXA; prothrombinase; Stuart factor; Stuart-Prower factor
Gene Symbols F10
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Activated factor Xa heavy chain.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2159
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.