Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor 11 light chain Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989117
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9117 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9117 Supplier Bioss Supplier No. BS9502R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen Factor 11 light chain
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene F11
Gene Accession No. P03951
Gene Alias Coagulation factor XI; Coagulation factor XIa heavy chain; Coagulation factor XIa light chain; F11; F11a; Factor 11; Factor11; FXI; Plasma thromboplastin antecedent; PTA
Gene Symbols F11
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Coagulation factor XIa light chain.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2160
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.