Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Factor IX Antibody (014), DyLight 680, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289372FR
Description
Factor IX Monoclonal specifically detects Factor IX in Human samples. It is validated for Sandwich ELISA Detection.Specifications
Factor IX | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
2158 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
14 | |
Sandwich ELISA Detection | |
Christmas factor, coagulation factor IX, EC 3.4.21, EC 3.4.21.22, factor 9, FIX, HEMB, MGC129641, MGC129642, plasma thromboplastic component, Plasma thromboplastin component, PTC | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Factor IX (AAB59620.1; Met1-Thr461). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction