Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Factor IX Recombinant Rabbit Monoclonal Antibody (3G3)
GREENER_CHOICE

Catalog No. PIMA538396
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538396 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538396 Supplier Invitrogen™ Supplier No. MA538396
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody has been tested in direct-ELISA.

Coagulation factor IX is a single chain polypeptide with a molecular mass of 57 kDa. Normal concentrations of factor IX in human plasma is approximately 3 μg/mL. Factor IX deficiency and lack of coagulation activity is seen in patients suffering from the innate X-chromosome linked disorder, haemophilia B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor IX
Applications Western Blot
Classification Recombinant Monoclonal
Clone 3G3
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 150 mM NaCl and 0.02% sodium azide; pH 7.4
Gene F9
Gene Accession No. P00740
Gene Alias AW111646; Cf9; Cf-9; Christmas factor; coagulation factor 9; coagulation factor IX; coagulation factor IX (plasma thromboplastic component, Christmas disease, hemophilia B); Coagulation factor IXa heavy chain; Coagulation factor IXa light chain; EC 3.4.21.22; F9; F9 p22; factor 9; factor IX (Christmas factor) precursor; factor IX F9; Factor9; FIX; FIX F9; HEMB; P19; plasma thromboplastic component; plasma thromboplastin component; PTC; RP6-88D7.1; serine protease; THPH8
Gene Symbols F9
Host Species Rabbit
Immunogen A synthesized peptide derived from human Factor IX.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2158
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.