Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Factor IX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA143067
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA143067 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA143067 Supplier Invitrogen™ Supplier No. PA143067
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

PA1-43067 detects Factor IX in rat, human, mouse and bovine samples. PA1-43067 has successfully been used in Western blot with human, mouse, bovine and rat samples. PA1-43067 has been used in ELISA applications with mouse and rat samples. PA1-43067 was derived from mouse factor IX.

Coagulation factor IX is a single chain polypeptide with a molecular mass of 57 kDa. Normal concentrations of factor IX in human plasma is approximately 3 μg/mL. Factor IX deficiency and lack of coagulation activity is seen in patients suffering from the innate X-chromosome linked disorder, haemophilia B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor IX
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 12.4 mg/mL
Conjugate Unconjugated
Formulation 50% water with 50% glycerol and no preservative
Gene F9
Gene Accession No. P00740, P00741, P16294, P16296
Gene Alias AW111646; Cf9; Cf-9; Christmas factor; coagulation factor 9; coagulation factor IX; coagulation factor IX (plasma thromboplastic component, Christmas disease, hemophilia B); Coagulation factor IXa heavy chain; Coagulation factor IXa light chain; EC 3.4.21.22; F9; F9 p22; factor 9; factor IX (Christmas factor) precursor; factor IX F9; Factor9; FIX; FIX F9; HEMB; P19; plasma thromboplastic component; plasma thromboplastin component; PTC; RP6-88D7.1; serine protease; THPH8
Gene Symbols F9
Host Species Sheep
Immunogen Rat Factor IX.
Purification Method SDS-PAGE
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14071, 2158, 24946, 280688
Target Species Human, Mouse, Rat, Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.