Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor IX Polyclonal Antibody, Invitrogen™

Catalog No. PIPA580608
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580608 Supplier Thermo Scientific Supplier No. PA580608

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Coagulation Factor IX/F9.

Factor IX Polyclonal antibody specifically detects Factor IX in Human samples. It is validated for ELISA, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor IX
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene F9
Gene Accession No. P00740
Gene Alias AW111646; Cf9; Cf-9; Christmas factor; coagulation factor 9; coagulation factor IX; coagulation factor IX (plasma thromboplastic component, Christmas disease, hemophilia B); Coagulation factor IXa heavy chain; Coagulation factor IXa light chain; EC 3.4.21.22; F9; F9 p22; factor 9; factor IX (Christmas factor) precursor; factor IX F9; Factor9; FIX; FIX F9; HEMB; P19; plasma thromboplastic component; plasma thromboplastin component; PTC; RP6-88D7.1; serine protease; THPH8
Gene Symbols F9
Host Species Rabbit
Immunogen Recombinant Human Coagulation Factor IX/F9 protein (Met1-Thr461)
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2158
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.