Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Factor V Light chain Monoclonal Antibody (B38)
GREENER_CHOICE

Catalog No. 01671854
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-854 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-854 Supplier Invitrogen™ Supplier No. MA110826
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10826 detects Factor V, light chain, from human samples. MA1-10826 has been successfully used in ELISA and immunohistochemistry applications. The MA1-10826 immunogen is purified human Factor V.

This gene encodes an essential cofactor of the blood coagulation cascade. This factor circulates in plasma, and is converted to the active form by the release of the activation peptide by thrombin during coagulation. This generates a heavy chain and a light chain which are held together by calcium ions. The activated protein is a cofactor that participates with activated coagulation factor X to activate prothrombin to thrombin. Defects in this gene result in either an autosomal recessive hemorrhagic diathesis or an autosomal dominant form of thrombophilia, which is known as activated protein C resistance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor V Light chain
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone B38
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene F5
Gene Accession No. P12259
Gene Alias Activated protein C cofactor; coagulation factor V; coagulation factor V (proaccelerin, labile factor); Coagulation factor V heavy chain; coagulation factor V jinjiang A2 domain; Coagulation factor V light chain; F5; factor V Leiden; FVL; PCCF; Proaccelerin, labile factor; RPRGL1; THPH2
Gene Symbols F5
Host Species Mouse
Immunogen Purified human Factor V.
Purification Method Purified
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2153
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.