Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Factor V Monoclonal Antibody (5104)
GREENER_CHOICE

Catalog No. PIMA143007
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA143007 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA143007 Supplier Invitrogen™ Supplier No. MA143007
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-43007 detects Factor V in bovine samples. MA1-43007 has successfully been used in RIA, WB, ELISA, IA applications. MA1-43007 was derived from bovine factor Va heavy chain.

Coagulation factor V (or Proaccelerin) is an essential factor of the blood coagulation cascade. This factor circulates in plasma, and is converted to the active form by the release of the activation peptide by thrombin during coagulation. This generates a heavy chain and a light chain which are held together by calcium ions. The active factor V is a cofactor that participates with activated coagulation factor X to activate prothrombin to thrombin. Defects in the Factor V gene can result various diseases including Factor V deficiency, thrombophilia due to activated protein C resistance, Budd-Chiari syndrome, Ischemic stroke, and recurrent pregnancy loss.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor V
Applications ELISA, Inhibition Assays, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone 5104
Concentration 9.5 mg/mL
Conjugate Unconjugated
Formulation 50% water with 50% glycerol and no preservative
Gene F5
Gene Accession No. Q28107
Gene Alias Ac2-120; Activated protein C cofactor; AI173222; Cf5; Cf-5; coagulation factor V; coagulation factor V (proaccelerin, labile factor); Coagulation factor V heavy chain; coagulation factor V jinjiang A2 domain; Coagulation factor V light chain; F5; Factor 5; factor V; factor V Leiden; Factor5; FVL; PCCF; Proaccelerin, labile factor; RPRGL1; THPH2
Gene Symbols F5
Host Species Mouse
Immunogen Bovine factor Va heavy chain.
Purification Method SDS-PAGE
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 280687
Target Species Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.