Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Factor V Monoclonal Antibody (5108)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA143002
Description
MA1-43002 detects Factor V in human samples. MA1-43002 has successfully been used in RIA and Western blot applications. MA1-43002 was derived from human factor V.
Coagulation factor V (or Proaccelerin) is an essential factor of the blood coagulation cascade. This factor circulates in plasma, and is converted to the active form by the release of the activation peptide by thrombin during coagulation. This generates a heavy chain and a light chain which are held together by calcium ions. The active factor V is a cofactor that participates with activated coagulation factor X to activate prothrombin to thrombin. Defects in the Factor V gene can result various diseases including Factor V deficiency, thrombophilia due to activated protein C resistance, Budd-Chiari syndrome, Ischemic stroke, and recurrent pregnancy loss.
Specifications
Factor V | |
Monoclonal | |
9.9 mg/mL | |
50% water with 50% glycerol and no preservative | |
P12259 | |
F5 | |
Human factor V and light chain (fragment E, 74,000) of factor Va. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Radioimmune Assays (RIA), Western Blot | |
5108 | |
Unconjugated | |
F5 | |
Ac2-120; Activated protein C cofactor; AI173222; Cf5; Cf-5; coagulation factor V; coagulation factor V (proaccelerin, labile factor); Coagulation factor V heavy chain; coagulation factor V jinjiang A2 domain; Coagulation factor V light chain; F5; Factor 5; factor V; factor V Leiden; Factor5; FVL; PCCF; Proaccelerin, labile factor; RPRGL1; THPH2 | |
Mouse | |
SDS-PAGE | |
RUO | |
2153 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction