Learn More
Invitrogen™ Factor V Polyclonal Antibody

Description
PA1-43041 detects Factor V in human samples. PA1-43041 has successfully been used in Western blot and ELISA applications. PA1-43041 was derived from human factor V.
Specifications
Specifications
| Antigen | Factor V |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 9.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50% water with 50% glycerol and no preservative |
| Gene | F5 |
| Gene Accession No. | P12259 |
| Gene Alias | Ac2-120; Activated protein C cofactor; AI173222; Cf5; Cf-5; coagulation factor V; coagulation factor V (proaccelerin, labile factor); Coagulation factor V heavy chain; coagulation factor V jinjiang A2 domain; Coagulation factor V light chain; F5; Factor 5; factor V; factor V Leiden; Factor5; FVL; PCCF; Proaccelerin, labile factor; RPRGL1; THPH2 |
| Gene Symbols | F5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.