Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Factor VII Monoclonal Antibody (AA-3)
GREENER_CHOICE

Catalog No. PIMA517633
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517633 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517633 Supplier Invitrogen™ Supplier No. MA517633
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17633 targets Factor VII/VIIa in ELISA and WB applications and shows reactivity with Human samples. The MA5-17633 immunogen is human Factor VII (AA-3) purified from plasma.

Coagulation factor VII is a vitamin K-dependent factor essential for hemostasis. This factor circulates in the blood in a zymogen form, and is converted to an active form by either factor IXa, factor Xa, factor XIIa, or thrombin by minor proteolysis. Upon activation of the factor VII, a heavy chain containing a catalytic domain and a light chain containing 2 EGF-like domains are generated, and two chains are held together by a disulfide bond. In the presence of factor III and calcium ions, the activated factor then further activates the coagulation cascade by converting factor IX to factor IXa and/or factor X to factor Xa. Defects in F7 production can cause coagulopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor VII
Applications ELISA, Western Blot
Classification Monoclonal
Clone AA-3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene F7
Gene Accession No. P08709
Gene Alias AI132620; Cf7; Coagulation factor VII; coagulation factor VII (serum prothrombin conversion accelerator); eptacog alfa; F7; Factor 7; Factor VII heavy chain; Factor VII light chain; Factor7; FVII; FVII coagulation protein; proconvertin; Serum prothrombin conversion accelerator; SPCA
Gene Symbols F7
Host Species Mouse
Immunogen Human Factor VII (AA-3) purified from plasma.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2155
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.