Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Factor VIII Antibody (402), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290255PCP
Description
Factor VIII Monoclonal antibody specifically detects Factor VIII in Human samples. It is validated for ELISASpecifications
Factor VIII | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
402 | |
ELISA | |
AHF, Antihemophilic factor, coagulation factor VIII, procoagulant component, coagulation factor VIIIc, DXS1253E, F8Ccoagulation factor VIII, factor VIII F8B, FVIII, HEMAF8B, Procoagulant component | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Factor VIII | |
0.1 mL | |
Cardiovascular Biology | |
2157 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction