Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Factor VIII:c heavy chain Antibody (RFF-VIII:c/10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP253119
Description
Factor VIII:c heavy chain Monoclonal specifically detects Factor VIII:c heavy chain in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Factor VIII:c heavy chain | |
Monoclonal | |
1.0 mg/mL | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, Immunocytochemistry, Immunofluorescence | |
RFF-VIII:c/10 | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
Purified human factor VIII | |
0.1 mg | |
Cardiovascular Biology | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction