Learn More
Factor VIII Monoclonal Antibody (F8 2.2.9), Invitrogen™
Description
MA1-10587 detects the fulll length of Factor VIII from human samples. This antibody does not cross-react with von Willebrand factor. MA1-10587 has been successfully used in ELISA and Western blot applications. The MA1-10587 immunogen is purified human Factor VIII.
Specifications
Specifications
| Antigen | Factor VIII |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | F8 2.2.9 |
| Concentration | 1.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | F8 |
| Gene Accession No. | P00451 |
| Gene Alias | AHF; Antihemophilic factor; CF8; Cf-8; coagulation factor VIII; coagulation factor VIII A1 domain; coagulation factor VIII C2 domain; coagulation factor VIII, procoagulant component; coagulation factor VIII, procoagulant component (hemophilia A); coagulation factor VIIIc; DXS1253E; F8; F8B; F8C; Factor 8; Factor VIII; Factor VIII B chain; factor VIII F8B; factor VIII precursor; Factor VIIIa heavy chain, 200 kDa isoform; Factor VIIIa heavy chain, 92 kDa isoform; Factor VIIIa light chain; factor8; FVIII; HEMA; Procoagulant component; RP11-115M6.7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.