Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Factor VIII Monoclonal Antibody (F8 2.2.9), Invitrogen™

Catalog No. PIMA546051
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546051 100 μg
PIMA545980 1 mg
01-671-541 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PIMA546051 Supplier Invitrogen™ Supplier No. MA546051
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10587 detects the fulll length of Factor VIII from human samples. This antibody does not cross-react with von Willebrand factor. MA1-10587 has been successfully used in ELISA and Western blot applications. The MA1-10587 immunogen is purified human Factor VIII.

This gene encodes coagulation factor VIII, which participates in the intrinsic pathway of blood coagulation; factor VIII is a cofactor for factor IXa which, in the presence of Ca+2 and phospholipids, converts factor X to the activated form Xa. This gene produces two alternatively spliced transcripts. Transcript variant 1 encodes a large glycoprotein, isoform a, which circulates in plasma and associates with von Willebrand factor in a noncovalent complex. This protein undergoes multiple cleavage events. Transcript variant 2 encodes a putative small protein, isoform b, which consists primarily of the phospholipid binding domain of factor VIIIc. This binding domain is essential for coagulant activity. Defects in this gene results in hemophilia A, a common recessive X-linked coagulation disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor VIII
Applications ELISA, Western Blot
Classification Monoclonal
Clone F8 2.2.9
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene F8
Gene Accession No. P00451
Gene Alias AHF; Antihemophilic factor; CF8; Cf-8; coagulation factor VIII; coagulation factor VIII A1 domain; coagulation factor VIII C2 domain; coagulation factor VIII, procoagulant component; coagulation factor VIII, procoagulant component (hemophilia A); coagulation factor VIIIc; DXS1253E; F8; F8B; F8C; Factor 8; Factor VIII; Factor VIII B chain; factor VIII F8B; factor VIII precursor; Factor VIIIa heavy chain, 200 kDa isoform; Factor VIIIa heavy chain, 92 kDa isoform; Factor VIIIa light chain; factor8; FVIII; HEMA; Procoagulant component; RP11-115M6.7
Gene Symbols F8
Host Species Mouse
Immunogen Purified human Factor VIII.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2157
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.