Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor VIII Mouse anti-Human, Clone: RFF-VIIIC/10, Novus Biologicals™
SDP

Catalog No. p-7201078 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018517 0.025 mg
NBP14102101 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018517 Supplier Novus Biologicals Supplier No. NBP1410210.025MG

Mouse Monoclonal Antibody

Factor VIII Monoclonal specifically detects Factor VIII in Human samples. It is validated for Western Blot, ELISA, Radioimmunoassay.

Specifications

Antigen Factor VIII
Applications ELISA, Western Blot, Radioimmune Assays (RIA)
Classification Monoclonal
Clone RFF-VIIIC/10
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100, ELISA 1:100-1:2000, Radioimmunoassay 1:100-1:2000
Formulation PBS (pH 7.4) with 0.09% Sodium Azide
Gene Accession No. P00451
Gene Alias AHF, Antihemophilic factor, coagulation factor VIII, procoagulant component, coagulation factor VIIIc, DXS1253E, F8Ccoagulation factor VIII, factor VIII F8B, FVIII, HEMAF8B, Procoagulant component
Gene Symbols F8
Host Species Mouse
Immunogen Affinity purified human Factor VIII
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2157
Test Specificity This antibody recognises human Factor VIII, an essential blood coagulation factor. Whilst circulating in the blood, it is mostly stably complexed to von Willebrand factor. It is activated through cleavage at various sites, dissociates from the complex and interacts with Factor IXa, in the presence of calcium ions and phospholipids, to convert Factor X to the activated Factor Xa, which activates thrombin. Thrombin cleaves fibrinogen into fibrin, which polymerises and cross-links to form a blood clot. The activated Factor VIII is proteolytically inactivated and cleared from the bloodstream. Clone RFF-VIIIC/10 recognises an epitope within the non-functional, middle domain of full-length Factor VIII.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.