Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor VIII Related Antigen/von Willebrand Factor Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS722P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
Prediluted
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS722P1 500 μL Concentrated 200μg/mL
MS722P1ABX 100 μL Concentrated 1mg/mL
MS722PABX 200 μL Concentrated 1mg/mL
MS722P 1 mL Concentrated 200μg/mL
MS722P0 100 μL Concentrated 200μg/mL
MS722R1 1 mL Prediluted Prediluted
MS722RQ 12 mL Ready-to-Use Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS722P1 Supplier Epredia Supplier No. MS722P1

Ensure accurate, reproducible results in immunohistochemistry experiments.

Factor VIII related antigen or von Willebrand factor is a multimeric glycoprotein, which binds to platelet glycoprotein Ib, glycoprotein IIb/IIIa, collagen and heparin. It is synthesized by endothelial cells and stored in the Weibel-Palade granules. It mediates platelet adhesion to injured vessel walls and serves as a carrier and stabilizer for coagulation factor VIII. von Willebrand factor is one of the most useful markers to identify endothelial (or megakaryocytic) lineage of neoplasms. As not all endothelial cells synthesize/store this molecule, about 30% of tumors of vascular origin fail to stain for factor VIII related antigen, regardless of whether they are benign or malignant. Staining for factor VIII related antigen has also been used to measure angiogenesis, an indicator of tumor recurrence.

Host Species: Mouse

Clone: F8/86

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Partially purified von Willebrand factor from human placenta

Molecular Weight: 270 kDa

Positive Control: Blood vessels in tonsil or intestine

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Factor VIII Related Antigen/von Willebrand Factor Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone F8/86
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Partially purified von Willebrand factor from human placenta
Quantity 500 μL
Regulatory Status IVD
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.