Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Factor X/Xa Monoclonal Antibody (5050)
GREENER_CHOICE

Catalog No. MA143013
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA143013 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA143013 Supplier Invitrogen™ Supplier No. MA143013
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody detects the heavy chains of human factor Xa and X; does not bind bovine factor X or BEGRck-Xa. Does not recognize murine or porcine X via Western blot. Inhibitory properties: inhibits PT, prothrombinase and partially APTT, but not amidase activity. Purity >95% by SDS-PAGE.

Factor X undergoes multiple processing steps before its preproprotein is converted to a mature two-chain form by the excision of the tripeptide RKR. Two chains of the factor are held together by 1 or more disulfide bonds; the light chain contains 2 EGF-like domains, while the heavy chain contains the catalytic domain which is structurally homologous to those of the other hemostatic serine proteases. The mature factor is activated by the cleavage of the activation peptide by factor IXa (in the intrisic pathway), or by factor VIIa (in the extrinsic pathway). The activated factor then converts prothrombin to thrombin in the presence of factor Va, Ca+2, and phospholipid during blood clotting. Mutations of this gene result in factor X deficiency, a hemorrhagic condition of variable severity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor X/Xa
Applications Affinity Purification, Inhibition Assays, Inhibition Assays, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone 5050
Concentration 7.6 mg/mL
Conjugate Unconjugated
Formulation 50% water with 50% glycerol and no preservative
Gene F10
Gene Accession No. P00742
Gene Alias Activated factor Xa heavy chain; blood coagulation factor X; Cf10; coagulation factor 10; coagulation factor X; coagulation factor X protein; F10; Factor X heavy chain; Factor X light chain; factor Xa; FX; FXA; prothrombinase; stuart factor; Stuart-Prower factor; Unknown (protein for IMAGE:8121240); unnamed protein product
Gene Symbols F10
Host Species Mouse
Immunogen Purified human Factor X.
Purification Method SDS-PAGE
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2159
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.