Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Factor X Recombinant Rabbit Monoclonal Antibody (3F2)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA549874
Description
Factor X Recombinant Monoclonal Antibody for IHC (P), ELISA
F10 undergoes multiple processing steps before its preproprotein is converted to a mature two-chain form by the excision of the tripeptide RKR. Two chains of the factor are held together by 1 or more disulfide bonds; the light chain contains 2 EGF-like domains, while the heavy chain contains the catalytic domain which is structurally homologous to those of the other hemostatic serine proteases. The mature factor is activated by the cleavage of the activation peptide by factor IXa (in the intrisic pathway), or by factor VIIa (in the extrinsic pathway). The activated factor then converts prothrombin to thrombin in the presence of factor Va, Ca+2, and phospholipid during blood clotting. Mutations of this gene result in factor X deficiency, a hemorrhagic condition of variable severity.
Specifications
Factor X | |
Recombinant Monoclonal | |
0.9 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P00742 | |
F10 | |
A synthesized peptide derived from human Factor X. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
3F2 | |
Unconjugated | |
F10 | |
Activated factor Xa heavy chain; blood coagulation factor X; Cf10; coagulation factor 10; coagulation factor X; coagulation factor X protein; F10; Factor 10; Factor X heavy chain; Factor X light chain; factor Xa; Factor10; FX; FXA; prothrombinase; RP11-98F14.9; stuart factor; Stuart-Prower factor; Unknown (protein for IMAGE:8121240); unnamed protein product | |
Rabbit | |
Affinity chromatography | |
RUO | |
2159 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction