Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor XII Antibody, Novus Biologicals™
SDP

Catalog No. NBP194203 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP194203 0.1 mL
NBP194203SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP194203 Supplier Novus Biologicals Supplier No. NBP194203
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Factor XII Polyclonal specifically detects Factor XII in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Factor XII
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:400
Formulation Whole antisera with 0.05% Sodium Azide
Gene Accession No. P00748
Gene Alias beta-factor XIIa part 1, coagulation factor XII, coagulation factor XII (Hageman factor), coagulation factor XIIa heavy chain, coagulation factor XIIa light chain, EC 3.4.21, EC 3.4.21.38, F12, HAE3, HAEX, HAF, HAF beta-factor XIIa part 2, Hageman factor
Gene Symbols F12
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human F12 protein (between residues 50-150) [UniProt P00748]
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2161
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.