Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Factor XII heavy chain Antibody (10-11-37), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP241377AF405
Description
Factor XII heavy chain Monoclonal specifically detects Factor XII heavy chain in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA.Specifications
Factor XII heavy chain | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
F12 | |
Affinity purified human factor XII. | |
0.1 mL | |
2161 | |
Human | |
Purified |
Western Blot, ELISA | |
13799 | |
Western Blot, ELISA, Sandwich ELISA | |
beta-factor XIIa part 1, coagulation factor XII, coagulation factor XII (Hageman factor), coagulation factor XIIa heavy chain, coagulation factor XIIa light chain, EC 3.4.21, EC 3.4.21.38, HAE3, HAEX, HAFbeta-factor XIIa part 2, Hageman factor | |
Mouse | |
Protein A or G purified | |
Primary | |
Antibody binds the heavy chain of human factor XII, including factor XII derivatives such as a-factor XIIa and a-factor XIIa C1-inhibitor complexes. | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction