Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor XII heavy chain Antibody (10-11-37), Alexa Fluor™ 594, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP241377AF594

Catalog No. NP241377A94


Only null left
Add to Cart

Description

Description

Factor XII heavy chain Monoclonal specifically detects Factor XII heavy chain in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA.
Specifications

Specifications

Factor XII heavy chain
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
F12
Affinity purified human factor XII.
Primary
Antibody binds the heavy chain of human factor XII, including factor XII derivatives such as a-factor XIIa and a-factor XIIa C1-inhibitor complexes.
Store at 4°C in the dark.
Western Blot, ELISA
13799
Western Blot, ELISA, Sandwich ELISA
beta-factor XIIa part 1, coagulation factor XII, coagulation factor XII (Hageman factor), coagulation factor XIIa heavy chain, coagulation factor XIIa light chain, EC 3.4.21, EC 3.4.21.38, HAE3, HAEX, HAFbeta-factor XIIa part 2, Hageman factor
Mouse
0.1 mL
2161
Human
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.