Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Factor XII heavy chain Antibody (10-11-5), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP241375MFV500
Description
Factor XII heavy chain Monoclonal antibody specifically detects Factor XII heavy chain in Human samples. It is validated for Western Blot, ELISA, ELISASpecifications
| Factor XII heavy chain | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 2161 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, ELISA | |
| 2005-10-11 00:00:00 | |
| Western Blot, ELISA, Sandwich ELISA | |
| beta-factor XIIa part 1, coagulation factor XII, coagulation factor XII (Hageman factor), coagulation factor XIIa heavy chain, coagulation factor XIIa light chain, EC 3.4.21, EC 3.4.21.38, HAE3, HAEX, HAFbeta-factor XIIa part 2, Hageman factor | |
| Affinity purified human factor XII. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction