Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Factor XII Heavy chain Monoclonal Antibody (B7C9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA121551
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Hemostasis following tissue injury involves the deployment of essential plasma procoagulants which are involved in a blood coagulation cascade leading to the formation of insoluble fibrin clots and the promotion of platelet aggregation. Factor XII, (FXII) a blood coagulation factor, is a serum glycoprotein that participates in fibrinolysis, as well as the generation of bradykinin and angiotensin. An enzyme of the serine protease (or serine endopeptidase) class, it activates both Factor XI and prekallikrein in the coagulation cascade. Factor XII deficiency, a rare hereditary disorder slightly more prevalent among Asians, does not cause excessive hemorrhaging since other coagulation factors compensate for it. Researchers have still reported Factor XII deficiency to be a risk factor for the development of arterial and venous thromboembolism. The gene for human Factor XII maps to the very end of the long arm of the fifth chromosome (5q33-qter).
Specifications
Factor XII Heavy chain | |
Monoclonal | |
1.4 mg/mL | |
PBS with no preservative; pH 7.4 | |
P00748 | |
F12 | |
Purified human Factor XII. | |
125 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
B7C9 | |
Unconjugated | |
F12 | |
Beta-factor XIIa part 1; beta-factor XIIa part 2; Coagulation factor XII; coagulation factor XII (Hageman factor); coagulation factor XIIa heavy chain; Coagulation factor XIIa light chain; F12; Factor 12; factor XII; Factor12; FXII; HAE3; HAEX; HAF; Hageman factor | |
Mouse | |
Protein G | |
RUO | |
2161 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction