Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Factor XII Heavy chain Monoclonal Antibody (B7C9)
GREENER_CHOICE

Catalog No. 01671856
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-856 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-856 Supplier Invitrogen™ Supplier No. MA110828
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10828 detects Factor XII heavy chain, from human samples. MA1-10828 has been successfully used in ELISA and Western blot applications. By Western blot, this antibody detects a ~50 kDa band representing the non-reduced heavy chain of Factor XII from normal human plasma . The MA1-10828 immunogen is purified human Factor XII.

Hemostasis following tissue injury involves the deployment of essential plasma procoagulants which are involved in a blood coagulation cascade leading to the formation of insoluble fibrin clots and the promotion of platelet aggregation. Factor XII, (FXII) a blood coagulation factor, is a serum glycoprotein that participates in fibrinolysis, as well as the generation of bradykinin and angiotensin. An enzyme of the serine protease (or serine endopeptidase) class, it activates both Factor XI and prekallikrein in the coagulation cascade. Factor XII deficiency, a rare hereditary disorder slightly more prevalent among Asians, does not cause excessive hemorrhaging since other coagulation factors compensate for it. Researchers have still reported Factor XII deficiency to be a risk factor for the development of arterial and venous thromboembolism. The gene for human Factor XII maps to the very end of the long arm of the fifth chromosome (5q33-qter).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor XII Heavy chain
Applications ELISA, Western Blot
Classification Monoclonal
Clone B7C9
Concentration 1.8 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene F12
Gene Accession No. P00748
Gene Alias Beta-factor XIIa part 1; beta-factor XIIa part 2; Coagulation factor XII; coagulation factor XII (Hageman factor); coagulation factor XIIa heavy chain; Coagulation factor XIIa light chain; F12; Factor 12; factor XII; Factor12; FXII; HAE3; HAEX; HAF; Hageman factor
Gene Symbols F12
Host Species Mouse
Immunogen Purified human Factor XII.
Purification Method Protein G
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2161
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.