Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Factor XII heavy chain Mouse anti-Human, Clone: 10.11.37, Novus Biologicals™

Mouse Monoclonal Antibody
$243.50
Specifications
Antigen | Factor XII heavy chain |
---|---|
Clone | 10.11.37 |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Factor XII heavy chain Monoclonal specifically detects Factor XII heavy chain in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA, Sandwich ELISA Capture.Specifications
Factor XII heavy chain | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Human | |
2161 | |
IgG2a κ | |
Protein A or G purified |
10.11.37 | |
Monoclonal | |
Purified | |
RUO | |
beta-factor XIIa part 1, coagulation factor XII, coagulation factor XII (Hageman factor), coagulation factor XIIa heavy chain, coagulation factor XIIa light chain, EC 3.4.21, EC 3.4.21.38, HAE3, HAEX, HAFbeta-factor XIIa part 2, Hageman factor | |
Affinity purified human factor XII. | |
Primary | |
Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title