Learn More
Invitrogen™ Factor XII Light chain Monoclonal Antibody (C6B7)

Description
MA1-10829 detects Factor XII, light chain, from human samples. MA1-10829 has been successfully used in ELISA applications. The MA1-10829 immunogen is purified human Factor XII.
Specifications
Specifications
| Antigen | Factor XII Light chain |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | C6B7 |
| Concentration | 3.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | F12 |
| Gene Accession No. | P00748 |
| Gene Alias | Beta-factor XIIa part 1; beta-factor XIIa part 2; Coagulation factor XII; coagulation factor XII (Hageman factor); coagulation factor XIIa heavy chain; Coagulation factor XIIa light chain; F12; Factor 12; Factor12; FXII; HAE3; HAEX; HAF; Hageman factor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.