Learn More
Epredia™ Lab Vision™ Factor XIII A Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Factor XIII A Ab-1, Mouse Monoclonal Antibody.
Supplier: Epredia™ MS1237R1
Description
Detect factor XIII in both reduced and non-reduced forms. This abtibody does not react with human Factor XIII B-chain or human Factor XII. Factor XIII is a Β-globulin found in plasma and is composed of two subunits. Factor XIII-A is the catalytic subunit and is a dimer of M.W. 160 kDa. Factor XIII is present in plasma as an alpha2Β2 heterodimer (M.W. 320 kDa); whereas in platelets, only the α2 unit exists. Factor XIIIa is a dermal dendrocyte marker and shows variable reaction with these types of tumors. It can be used for histiocytic phenotyping and has been reported to mark capillary hemangiomas and tumors of the central nervous system. Factor XIII has also been used with CD34 to differentiate between dermatofibroma and dermatofibrosarcoma protuberans.Host Species: Mouse
Clone: AC-1A1
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant protein corresponding to A-subunit of coagulation Factor XIII
Molecular Weight: 160 kDa (unreduced dimmer); 80 kDa (reduced monomer)
Positive Control: Recombinant human Factor XIII A-subunit; Capillary hemangioma, dermatofibroma and placenta
Cellular Localization: Cytoplasmic and nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
Factor XIII A Ab-1 | |
Monoclonal | |
— | |
Ready-to-use for IHC (UVLP) | |
Recombinant protein corresponding to A-subunit of coagulation Factor XIII | |
IVD | |
Primary | |
Prediluted |
Immunohistochemistry (Paraffin), Western Blot | |
AC-1A1 | |
Unconjugated | |
Mouse | |
1 mL | |
Hematopoietic | |
Human | |
IgG2a κ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.