Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Factor XIII A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522110
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522110 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522110 Supplier Invitrogen™ Supplier No. PA522110
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: H1299. Predicted reactivity: Mouse (86%), Rat (84%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes the coagulation factor XIII A subunit. Coagulation factor XIII is the last zymogen to become activated in the blood coagulation cascade. Plasma factor XIII is a heterotetramer composed of 2 A subunits and 2 B subunits. The A subunits have catalytic function, and the B subunits do not have enzymatic activity and may serve as plasma carrier molecules. Platelet factor XIII is comprised only of 2 A subunits, which are identical to those of plasma origin. Upon cleavage of the activation peptide by thrombin and in the presence of calcium ion, the plasma factor XIII dissociates its B subunits and yields the same active enzyme, factor XIIIa, as platelet factor XIII. This enzyme acts as a transglutaminase to catalyze the formation of gamma-glutamyl-epsilon-lysine crosslinking between fibrin molecules, thus stabilizing the fibrin clot. It also crosslinks alpha-2-plasmin inhibitor, or fibronectin, to the alpha chains of fibrin. Factor XIII deficiency is classified into two categories: type I deficiency, characterized by the lack of both the A and B subunits; and type II deficiency, characterized by the lack of the A subunit alone. These defects can result in a lifelong bleeding tendency, defective wound healing, and habitual abortion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor XIII A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene F13A1
Gene Accession No. P00488
Gene Alias 1200014I03Rik; AI462306; bA525O21.1 (coagulation factor XIII, A1 polypeptide); coagulation factor XIII A chain; coagulation factor XIII, A polypeptide; coagulation factor XIII, A1 polypeptide; coagulation factor XIII, A1 subunit; coagulation factor XIII, alpha subunit; coagulation factor XIIIa; F13A; F13a1; factor viiia; Factor XIIIA; fibrin stabilizing factor, A subunit; fibrinoligase; FSF, A subunit; protein-glutamine gamma-glutamyltransferase A chain; RP11-232H4.1; TGase; transglutaminase A chain; transglutaminase. plasma
Gene Symbols F13A1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human Factor XIIIa. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2162
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.