Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Factor XIII B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552004
Description
Immunogen sequence: FCLAGYTTES GRQEEQTTCT TEGWSPEPRC FKKCTKPDLS NGYISDVKLL YKIQENMHYG CASGYKTTGG KDEEVVQCLS DGWSSQPTCR KEHETCLAPE LYNGNYSTTQ KTFKVKDKVQ YECATGYYTA GGKKTEEV Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 79%.
This gene encodes coagulation factor XIII B subunit. Coagulation factor XIII is the last zymogen to become activated in the blood coagulation cascade. Plasma factor XIII is a heterotetramer composed of 2 A subunits and 2 B subunits. The A subunits have catalytic function, and the B subunits do not have enzymatic activity and may serve as a plasma carrier molecules. Platelet factor XIII is comprised only of 2 A subunits, which are identical to those of plasma origin. Upon activation by the cleavage of the activation peptide by thrombin and in the presence of calcium ion, the plasma factor XIII dissociates its B subunits and yields the same active enzyme, factor XIIIa, as platelet factor XIII. This enzyme acts as a transglutaminase to catalyze the formation of gamma-glutamyl-epsilon-lysine crosslinking between fibrin molecules, thus stabilizing the fibrin clot. Factor XIII deficiency is classified into two categories: type I deficiency, characterized by the lack of both the A and B subunits; and type II deficiency, characterized by the lack of the A subunit alone. These defects can result in a lifelong bleeding tendency, defective wound healing, and habitual abortion.
Specifications
Factor XIII B | |
Polyclonal | |
Unconjugated | |
F13B | |
coagulation factor XIII B chain; coagulation factor XIII, B polypeptide; F13B; Fibrin-stabilizing factor B subunit; FXIIIB; Protein-glutamine gamma-glutamyltransferase B chain; TGase; transglutaminase B chain | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2165 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P05160 | |
F13B | |
Recombinant protein corresponding to Human Factor XIII B. Recombinant protein control fragment (Product #RP-100497). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction