Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Factor XIIIa Antibody (F13A1/3772R) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308933
Description
Factor XIIIa Monoclonal specifically detects Factor XIIIa in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Factor XIIIa | |
Monoclonal | |
1 mg/mL | |
Immunohistochemistry-Paraffin : 1-2 ug/mL | |
bA525O21.1 (coagulation factor XIII, A1 polypeptide), coagulation factor XIII A chain, coagulation factor XIII, A1 polypeptide, Coagulation factor XIIIa, EC 2.3.2.13, F13Acoagulation factor XIII, A polypeptide, factor XIIIa, fibrin stabilizing factor, A subunit, fibrinoligase, FSF, A subunit, Protein-glutamine gamma-glutamyltransferase A chain, TGase, Transglutaminase A chain, transglutaminase. plasma | |
Recombinant fragment of human Factor XIIIa protein (aa46-181) (exact sequence is proprietary) | |
100 μg | |
Apoptosis, Cancer, Cell Biology | |
2162 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry (Paraffin) | |
F13A1/3772R | |
Unconjugated | |
10mM PBS | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction