Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Factor XIIIb Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299806H
Description
Factor XIIIb Polyclonal specifically detects Factor XIIIb in Human samples. It is validated for ELISA.Specifications
Factor XIIIb | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
2165 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
HRP | |
coagulation factor XIII B chain, coagulation factor XIII, B polypeptide, F13B, fibrin-stabilizing factor B subunit, FXIIIB, protein-glutamine gamma-glutamyltransferase B chain, TGase, transglutaminase B chain | |
Produced in rabbits immunized with purified, recombinant Factor XIIIb (P05160; Met1-Thr661). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction