Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Factor XIIIb Antibody (OTI1A3), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270697H
Description
Factor XIIIb Monoclonal specifically detects Factor XIIIb in Human samples. It is validated for Western Blot.Specifications
Factor XIIIb | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
F13B | |
Recombinant protein fragment corresponding to amino acids 357-661 of human Factor XIII (NP_001985) produced in E.coli. | |
0.1 mL | |
2165.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1A3 | |
Western Blot | |
coagulation factor XIII B chain, coagulation factor XIII, B polypeptide, F13B, fibrin-stabilizing factor B subunit, FXIIIB, protein-glutamine gamma-glutamyltransferase B chain, TGase, transglutaminase B chain | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction