Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Factor XIIIb Antibody (OTI1A3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245563
Description
Ensure accurate, reproducible results in Western Blot
Factor XIIIb Monoclonal specifically detects Factor XIIIb in Human samples. It is validated for Western Blot.Specifications
Factor XIIIb | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 | |
coagulation factor XIII B chain, coagulation factor XIII, B polypeptide, F13B, fibrin-stabilizing factor B subunit, FXIIIB, protein-glutamine gamma-glutamyltransferase B chain, TGase, transglutaminase B chain | |
Mouse | |
73.2 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
OTI1A3 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
F13B | |
Human recombinant protein fragment corresponding to amino acids 357-661 of human F13B/Factor XIIIb (NP_001985) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
2165 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction