Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FADD Antibody (OTI1C11), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271387V
Description
FADD Monoclonal specifically detects FADD in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
FADD | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FADD | |
Full length recombinant protein of human FADD (NP_003815) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry | |
OTI1C11 | |
Western Blot, Flow Cytometry | |
Fas (TNFRSF6)-associated via death domain, FAS-associated death domain protein, FAS-associating death domain-containing protein, GIG3, Growth-inhibiting gene 3 protein, Mediator of receptor induced toxicity, mediator of receptor-induced toxicity, MGC8528, MORT1Fas-associating protein with death domain, protein FADD | |
Mouse | |
Affinity Purified | |
Apoptosis, Cancer, Death Receptor Signaling Pathway, Diabetes Research, Necroptosis, Phospho Specific, Tumor Suppressors | |
8772.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction