Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FADD Monoclonal Antibody (FD19)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA120168
Description
Recommended positive controls: A431. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is an adaptor molecule that interacts with various cell surface receptors and mediates cell apoptotic signals. Through its C-terminal death domain, this protein can be recruited by TNFRSF6/Fas-receptor, tumor necrosis factor receptor, TNFRSF25, and TNFSF10/TRAIL-receptor, and thus it participates in the death signaling initiated by these receptors. Interaction of this protein with the receptors unmasks the N-terminal effector domain of this protein, which allows it to recruit caspase-8, and thereby activate the cysteine protease cascade. Knockout studies in mice also suggest the importance of this protein in early T cell development.
Specifications
FADD | |
Monoclonal | |
2.1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
Q13158 | |
FADD | |
Recombinant, full length, human FADD protein. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
ELISA, Immunoprecipitation, Western Blot | |
FD19 | |
Unconjugated | |
FADD | |
Fadd; Fas (TNFRSF6)-associated via death domain; Fas associated via death domain; FAS-associated death domain protein; FAS-associating death domain-containing protein; Fas-associating protein with death domain; GIG3; Growth-inhibiting gene 3 protein; mediator of receptor induced toxicity; mediator of receptor-induced toxicity; MGC8528; MORT1; Mort1/FADD; protein FADD | |
Mouse | |
Purified | |
RUO | |
266610, 8772 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction